Learn More
Apexbio Technology LLC Coenzyme Q0 605-94-7 1g
SDP
Catalog No. 5000093353
Encompass_Preferred
Coenzyme Q0 (CAS 605-94-7) is a quinone derivative originally derived from cinnamon extract It modulates multiple cellular pathways by inducing apoptosis and autophagy primarily through inhibition of the HER-2/AKT/mTOR signaling cascade Additionally Coenzyme Q0 regulates transcription factors such as NF B and AP-1 enhances Nrf2 stability and mitigates inflammation and oxidative stress Its anti-angiogenic properties are mediated via downregulation of the MMP-9/NF- B pathway and upregulation of HO-1 signaling Coenzyme Q0 is widely used in basic research and drug development targeting cancer inflammation and angiogenesis-related disorders
Catalog No. 50-000-93353 Supplier Apexbio Technology LLC Supplier No. A47831000
Coenzyme Q0 (CAS 605-94-7) is a quinone derivative originally derived from cinnamon extract It modulates multiple cellular pathways by inducing apoptosis and autophagy primarily through inhibition of the HER-2/AKT/mTOR signaling cascade Additionally Coenzyme Q0 regulates transcription factors such as NF B and AP-1 enhances Nrf2 stability and mitigates inflammation and oxidative stress Its anti-angiogenic properties are mediated via downregulation of the MMP-9/NF- B pathway and upregulation of HO-1 signaling Coenzyme Q0 is widely used in basic research and drug development targeting cancer inflammation and angiogenesis-related disorders