Learn More
Cayman Chemical Coenzyme Q1 10mg
SDP
Catalog No. 502979418
Encompass_Preferred
An electron acceptor and a derivative of the mitochondrial electron transport chain cofactor CoQ10; induces the permeability transition pore opening and induces apoptosis in clone 9 cells at 50 µM; is used as a marker for mitochondrial complex I activity in perfused tissue and subcellular fractions
Catalog No. 50-297-9418 Supplier Cayman Chemical Supplier No. 1874110 MG
An electron acceptor and a derivative of the mitochondrial electron transport chain cofactor CoQ10; induces the permeability transition pore opening and induces apoptosis in clone 9 cells at 50 µM; is used as a marker for mitochondrial complex I activity in perfused tissue and subcellular fractions