Learn More
Cayman Chemical Coenzyme Q10 1G
SDP
Catalog No. 502954376
Encompass_Preferred
A cofactor in the electron transport chain; decreased levels due to genetic mutations in its synthesis enzymes lead to encephalopathy, cerebellar ataxia, infantile multisystemic form, nephropathy, and isolated myopathy
Catalog No. 50-295-4376 Supplier Cayman Chemical Supplier No. 115061 G
A cofactor in the electron transport chain; decreased levels due to genetic mutations in its synthesis enzymes lead to encephalopathy, cerebellar ataxia, infantile multisystemic form, nephropathy, and isolated myopathy