Learn More
Apexbio Technology LLC Coenzyme Q10 303-98-0 200mg
SDP
Catalog No. 5000090329
Encompass_Preferred
Coenzyme Q10 (CAS 303-98-0) is a lipophilic benzoquinone involved in mitochondrial electron transport and cellular energy production It functions as an essential electron carrier within the electron transport chain facilitating ATP synthesis and exhibiting antioxidant properties by mitigating oxidative damage Coenzyme Q10 is frequently studied for its role in cellular metabolism bioenergetics and redox homeostasis Due to its involvement in mitochondrial function it is widely applied in research on metabolic disorders neurodegeneration and cardiovascular diseases The compound is soluble in organic solvents such as chloroform benzene and carbon tetrachloride For optimal stability it should be stored sealed in cool and dry conditions
Catalog No. 50-000-90329 Supplier Apexbio Technology LLC Supplier No. N1796200
Coenzyme Q10 (CAS 303-98-0) is a lipophilic benzoquinone involved in mitochondrial electron transport and cellular energy production It functions as an essential electron carrier within the electron transport chain facilitating ATP synthesis and exhibiting antioxidant properties by mitigating oxidative damage Coenzyme Q10 is frequently studied for its role in cellular metabolism bioenergetics and redox homeostasis Due to its involvement in mitochondrial function it is widely applied in research on metabolic disorders neurodegeneration and cardiovascular diseases The compound is soluble in organic solvents such as chloroform benzene and carbon tetrachloride For optimal stability it should be stored sealed in cool and dry conditions