Learn More
Selleck Chemical LLC Coenzyme Q10 (CoQ10) 200mg 303-98-0
SDP
Catalog No. 506465
Encompass_Preferred
Coenzyme Q10 (ubiquinone, ubidecarenone, coenzyme Q) is a component of the electron transport chain and participates in aerobic cellular respiration. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-646-5 Supplier Selleck Chemical LLC Supplier No. S2398200MG
Coenzyme Q10 (ubiquinone, ubidecarenone, coenzyme Q) is a component of the electron transport chain and participates in aerobic cellular respiration. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.