Learn More
Cayman Chemical Coenzyme Q10-d6 1mg
SDP
Catalog No. 502989250
Encompass_Preferred
An internal standard for the quantification of CoQ10 by GC- or LC-MS
Catalog No. 50-298-9250 Supplier Cayman Chemical Supplier No. 309581 MG
An internal standard for the quantification of CoQ10 by GC- or LC-MS