Learn More
Medchemexpress LLC Coenzyme Q10-d9 | 2687960-97-8 | 98.2% | 872.40 | 1 MG
SDP
Catalog No. 5000428753
Encompass_Preferred
Coenzyme Q10-d9 is the deuterium labeled Coenzyme Q10. Coenzyme Q10 is an essential cofactor of the electron transport chain and a potent antioxidant agent.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-28753 Supplier Medchemexpress LLC Supplier No. HYN0111S21MG
Coenzyme Q10-d9 is the deuterium labeled Coenzyme Q10. Coenzyme Q10 is an essential cofactor of the electron transport chain and a potent antioxidant agent.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS