Learn More
Cayman Chemical Coenzyme Q2 5mg
SDP
Catalog No. 502979649
Encompass_Preferred
A biosynthetic precursor to CoQ10 and an inhibitor of mitochondrial complex I; an electron acceptor that accepts electrons from mitochondrial complex I; inhibits proliferation of BALL-1 leukemia cells (IC50 = 20 µM) and induces apoptosis in Huh7 and HepG2 cells at 100 µM; increases the production of ROS in BALL-1 cells but inhibits ROS production in mitochondrial membranes derived from bovine heart in combination with rotenone, piericidin A, or rolliniastatin 1 or -2; increases the production of hydrogen peroxide in isolated rabbit heart mitochondria and decreases it in isolated rabbit liver mitochondria
Catalog No. 50-297-9649 Supplier Cayman Chemical Supplier No. 173275 MG
A biosynthetic precursor to CoQ10 and an inhibitor of mitochondrial complex I; an electron acceptor that accepts electrons from mitochondrial complex I; inhibits proliferation of BALL-1 leukemia cells (IC50 = 20 µM) and induces apoptosis in Huh7 and HepG2 cells at 100 µM; increases the production of ROS in BALL-1 cells but inhibits ROS production in mitochondrial membranes derived from bovine heart in combination with rotenone, piericidin A, or rolliniastatin 1 or -2; increases the production of hydrogen peroxide in isolated rabbit heart mitochondria and decreases it in isolated rabbit liver mitochondria