Learn More
Cayman Chemical Coenzyme Q9 1mg
SDP
Catalog No. 502980396
Encompass_Preferred
A mitochondrial ubiquinone; reduces the incidence of ischemia and reperfusion-induced arrhythmia and improves post-ischemic heart rate, coronary flow, aortic flow, and left ventricular developed pressure in isolated guinea pig hearts at 5 mg/kg; reduces ischemia and reperfusion-induced myocardial infarct size and the number of apoptotic cardiomyocytes in the same model at 5 mg/kg
Catalog No. 50-298-0396 Supplier Cayman Chemical Supplier No. 168661 MG
A mitochondrial ubiquinone; reduces the incidence of ischemia and reperfusion-induced arrhythmia and improves post-ischemic heart rate, coronary flow, aortic flow, and left ventricular developed pressure in isolated guinea pig hearts at 5 mg/kg; reduces ischemia and reperfusion-induced myocardial infarct size and the number of apoptotic cardiomyocytes in the same model at 5 mg/kg