Learn More
Medchemexpress LLC HY-101415 1mg Medchemexpress, Coenzyme Q9 CAS:303-97-9 Purity:>98%
SDP
Catalog No. 501932117
Encompass_Preferred
Medchemexpress, HY-101415 1mg Coenzyme Q9 CAS:303-97-9 Coenzyme Q 9 , a nine isoprenyl group-containing member of the ubiquinone family, is a normal constituent of human plasma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2117 Supplier Medchemexpress LLC Supplier No. HY1014151MG
Medchemexpress, HY-101415 1mg Coenzyme Q9 CAS:303-97-9 Coenzyme Q 9 , a nine isoprenyl group-containing member of the ubiquinone family, is a normal constituent of human plasma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.