Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ COEX Cold Extractor
SDP

Catalog No. 11392216
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
11-392-216 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-216 Supplier Velp™ Supplier No. F30540204
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cold extractor that performs rapid fat extraction on feed samples directly in the FIWE Advance and FIWE glass crucibles.

  • Performs preliminary defatting of sample prior to fiber determination with FIWE and FIWE Advance
  • Guarantees maximum reliability and is a mandatory step for all feed sample with high fat content
  • Efficient pump aspirates and removes fatty component that will be collected in a dedicated waste tank or bottle
  • Easy to use and convenient, as all reagents can be added through the same channel
  • Solvent acts directly on the sample
  • Ensures rapid fat extraction directly into the same glass crucibles used by the FIWE and FIWE Advance whenever fat content of feed sample is high and needs to be reduced to less than 1%
  • Crucible containing defatted sample is transferred directly to the FIWE and FIWE Advance analyzer, eliminating possible errors due to sample transfer

Specifications

Product Type Cold Extractor
For Use With (Equipment) COEX Cold Extractor
Frequency 60 Hz
Voltage 115 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.