Learn More
Instech Co-extruded PE/PVC external infusion tubing. Polyethylene flow path for biocompatibility; PVC outside to minimize kinking. .024x.064in; fits 22ga. Clear. 30m (98ft) spool. Non sterile.
SDP
Catalog No. NC0614261
Encompass
The Instech ABS2/TS062-W5 is a sterile tubing set for the ABS2 automated blood sampler, providing waste and fluid-path segments for drawing blood, returning IV fluid, and routing samples to the cooled collector. It also serves as replacement waste tubing segments and vials for maintaining system performance across mouse, rat, and large-animal ABS2 setups. Designed for accurate, low-loss sampling in closed-system PK workflows.
Features
  • Sterile tubing set for ABS2 sampler
  • Includes waste-path segments and vials
  • Supports blood draw and IV return
  • Low-loss routing for undiluted samples
  • Compatible with multi-species ABS2 setups
Catalog No. NC0614261 Supplier Instech Supplier No. BTCOEX22
The Instech ABS2/TS062-W5 is a sterile tubing set for the ABS2 automated blood sampler, providing waste and fluid-path segments for drawing blood, returning IV fluid, and routing samples to the cooled collector. It also serves as replacement waste tubing segments and vials for maintaining system performance across mouse, rat, and large-animal ABS2 setups. Designed for accurate, low-loss sampling in closed-system PK workflows.
Features
  • Sterile tubing set for ABS2 sampler
  • Includes waste-path segments and vials
  • Supports blood draw and IV return
  • Low-loss routing for undiluted samples
  • Compatible with multi-species ABS2 setups