Learn More
Instech Co-extruded PE/PVC external infusion tubing. Polyethylene flow path for biocompatibility; PVC outside to minimize kinking. .017x.051in; fits 25ga. Clear. 30m (98ft) spool. Non sterile.
SDP
Catalog No. 50890047
Encompass
The Instech BTCOEX‑25 is a co‑extruded PE/PVC external infusion tubing featuring a polyethylene inner layer for excellent compound compatibility and a PVC outer layer to minimize kinking and evaporation. Sized at .017x.051in (0.4x1.3mm) to fit 25ga connectors, this clear tubing provides a reliable friction‑fit interface for external infusion lines. It is supplied non‑sterile on a 30m (98ft) spool for use in laboratory research applications.
Features
  • Co‑extruded PE/PVC external infusion tubing
  • PE inner layer for biocompatible flow path
  • PVC outer layer for kink resistance
  • .017x.051in (0.4x1.3mm) dimensions; fits 25ga
  • Clear tubing on a 30m (98ft) non‑sterile spool
  • Designed for external infusion use only
Catalog No. 50-890-047 Supplier Instech Supplier No. BTCOEX25
The Instech BTCOEX‑25 is a co‑extruded PE/PVC external infusion tubing featuring a polyethylene inner layer for excellent compound compatibility and a PVC outer layer to minimize kinking and evaporation. Sized at .017x.051in (0.4x1.3mm) to fit 25ga connectors, this clear tubing provides a reliable friction‑fit interface for external infusion lines. It is supplied non‑sterile on a 30m (98ft) spool for use in laboratory research applications.
Features
  • Co‑extruded PE/PVC external infusion tubing
  • PE inner layer for biocompatible flow path
  • PVC outer layer for kink resistance
  • .017x.051in (0.4x1.3mm) dimensions; fits 25ga
  • Clear tubing on a 30m (98ft) non‑sterile spool
  • Designed for external infusion use only