Learn More
MSC Scot's Tuff Coffee Spot Remover, Case of 12, 32 oz Bottles
Catalog No. 502838210
Encompass
Scot's Tuff Coffee Spot Remover from Scot Laboratories is a specialized cleaning solution designed for effective spot removal on carpets and upholstery. This product is supplied in a case of twelve 32 oz bottles. It is a clear, light yellow liquid with a bitter odor and a pH of 4.5, offering complete solubility in water. Users should note that it causes serious eye damage and is harmful to aquatic life.
  • Spot remover for carpet and upholstery care
  • Supplied in a case of 12 bottles
  • Each bottle contains 32 oz of cleaner
  • Clear liquid with a light yellow color
  • Bitter odor
  • pH of 4.5
  • Completely soluble in water
Catalog No. 50-283-8210 Supplier MSC Supplier No. 86131984
Scot's Tuff Coffee Spot Remover from Scot Laboratories is a specialized cleaning solution designed for effective spot removal on carpets and upholstery. This product is supplied in a case of twelve 32 oz bottles. It is a clear, light yellow liquid with a bitter odor and a pH of 4.5, offering complete solubility in water. Users should note that it causes serious eye damage and is harmful to aquatic life.
  • Spot remover for carpet and upholstery care
  • Supplied in a case of 12 bottles
  • Each bottle contains 32 oz of cleaner
  • Clear liquid with a light yellow color
  • Bitter odor
  • pH of 4.5
  • Completely soluble in water