Learn More
MSC Coffing Hand Chain Hoist, 1/2 ton Capacity, 10 ft Lift
Catalog No. 5000513812
Encompass
This hand chain hoist offers a 1/2 ton capacity and 10 ft lifting height. Its compact, lightweight design is ideal for construction and contractor applications, facilitating portability and easy rigging. Precise load handling is ensured by an instant-engaging mechanical load brake.
  • Compact, lightweight design for portability and easy rigging.
  • Lubricated bearings, bushings, and heat-treated spur gearing ensure smooth, efficient operation.
  • Mechanical load brake engages instantly for precise load holding and positioning.
  • 360-degree swivel hooks with safety latches ensure positive load engagement.
  • Durable through-hardened load chain provides added strength and wear resistance.
  • Stamped steel housing withstands rugged conditions.
  • Designed for easy inspection and maintenance, allowing simple hand tool dismantling.
  • Meets ANSI/ASME B30.16 standards.
Catalog No. 50-005-13812 Supplier MSC Supplier No. 65178238
This hand chain hoist offers a 1/2 ton capacity and 10 ft lifting height. Its compact, lightweight design is ideal for construction and contractor applications, facilitating portability and easy rigging. Precise load handling is ensured by an instant-engaging mechanical load brake.
  • Compact, lightweight design for portability and easy rigging.
  • Lubricated bearings, bushings, and heat-treated spur gearing ensure smooth, efficient operation.
  • Mechanical load brake engages instantly for precise load holding and positioning.
  • 360-degree swivel hooks with safety latches ensure positive load engagement.
  • Durable through-hardened load chain provides added strength and wear resistance.
  • Stamped steel housing withstands rugged conditions.
  • Designed for easy inspection and maintenance, allowing simple hand tool dismantling.
  • Meets ANSI/ASME B30.16 standards.