Learn More
ECM Bioscience Cofilin Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068621
Encompass_Preferred
The cofilin phospho-regulation antibody sampler kit can be used to monitor the level of phosphorylated cofilin relative to total cofilin expression. In addition, the kit provides reagents for examining the activity and expression levels of the cofilin kinase, LIMK1, and cofilin phosphatase, Slingshot. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms, Ck
Catalog No. 50-206-8621 Supplier ECM Bioscience Supplier No. CK6040
The cofilin phospho-regulation antibody sampler kit can be used to monitor the level of phosphorylated cofilin relative to total cofilin expression. In addition, the kit provides reagents for examining the activity and expression levels of the cofilin kinase, LIMK1, and cofilin phosphatase, Slingshot. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms, Ck