Learn More
Medchemexpress LLC COG 133 | 514200-66-9 | 98.7% | 2169.71 | 1 MG
SDP
Catalog No. 5000424548
Encompass_Preferred
COG 133 is a fragment of Apolipoprotein E (APOE) peptide. It competes with the ApoE holoprotein for binding the LDL receptor, showing potent anti-inflammatory and neuroprotective effects. It also acts as a nAChR antagonist with an IC50 of 445 nM.
  • Fragment of apolipoprotein E (APOE) peptide
  • Competes with ApoE holoprotein for LDL receptor binding
  • Shows potent anti-inflammatory effects
  • Provides neuroprotective benefits
  • Functions as a nAChR antagonist
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-004-24548 Supplier Medchemexpress LLC Supplier No. HYP10501MG
COG 133 is a fragment of Apolipoprotein E (APOE) peptide. It competes with the ApoE holoprotein for binding the LDL receptor, showing potent anti-inflammatory and neuroprotective effects. It also acts as a nAChR antagonist with an IC50 of 445 nM.
  • Fragment of apolipoprotein E (APOE) peptide
  • Competes with ApoE holoprotein for LDL receptor binding
  • Shows potent anti-inflammatory effects
  • Provides neuroprotective benefits
  • Functions as a nAChR antagonist
  • White to off-white solid appearance
  • For research use only