Learn More
Medchemexpress LLC Cog 133 | 514200-66-9 | 98.7% | 2169.71 | 10 MG
SDP
Catalog No. 5000424547
Encompass_Preferred
COG 133 is a fragment of Apolipoprotein E (APOE) peptide. It competes with the ApoE holoprotein for binding the LDL receptor and exhibits potent anti-inflammatory and neuroprotective effects. Additionally, it acts as a nAChR antagonist with an IC50 of 445 nM.
  • Fragment of Apolipoprotein E (APOE) peptide
  • Competes with ApoE holoprotein for LDL receptor binding
  • Potent anti-inflammatory effects
  • Neuroprotective effects
  • nAChR antagonist (IC50 of 445 nM)
Catalog No. 50-004-24547 Supplier Medchemexpress LLC Supplier No. HYP105010MG
COG 133 is a fragment of Apolipoprotein E (APOE) peptide. It competes with the ApoE holoprotein for binding the LDL receptor and exhibits potent anti-inflammatory and neuroprotective effects. Additionally, it acts as a nAChR antagonist with an IC50 of 445 nM.
  • Fragment of Apolipoprotein E (APOE) peptide
  • Competes with ApoE holoprotein for LDL receptor binding
  • Potent anti-inflammatory effects
  • Neuroprotective effects
  • nAChR antagonist (IC50 of 445 nM)