Learn More
Medchemexpress LLC COG 133 | 514200-66-9 | 98.7% | 2169.71 | 5 MG
SDP
Catalog No. 5000423868
Encompass_Preferred
COG 133 is a fragment of Apolipoprotein E (APOE) peptide. It competes with the ApoE holoprotein for binding the LDL receptor and acts as a nAChR antagonist with an IC50 of 445 nM.
  • Potent anti-inflammatory effects
  • Neuroprotective effects
  • nAChR antagonist with an IC50 of 445 nM
Catalog No. 50-004-23868 Supplier Medchemexpress LLC Supplier No. HYP10505MG
COG 133 is a fragment of Apolipoprotein E (APOE) peptide. It competes with the ApoE holoprotein for binding the LDL receptor and acts as a nAChR antagonist with an IC50 of 445 nM.
  • Potent anti-inflammatory effects
  • Neuroprotective effects
  • nAChR antagonist with an IC50 of 445 nM