Learn More
Apexbio Technology LLC COG 133 TFA 2828432-37-5 1mg
SDP
Catalog No. 5000093824
Encompass_Preferred
COG 133 TFA (CAS 2828432-37-5) is a peptide fragment derived from apolipoprotein E (APOE) that competitively binds to low-density lipoprotein receptors modulating associated signaling pathways This compound exhibits both anti-inflammatory and neuroprotective properties with an antagonist activity concentration of 445 nM reported in relevant assays Owing to its specific mechanism of action and demonstrated biological effects COG 133 TFA is widely utilized in research focused on neurodegenerative disorders and inflammation-related pathologies
Catalog No. 50-000-93824 Supplier Apexbio Technology LLC Supplier No. A89611
COG 133 TFA (CAS 2828432-37-5) is a peptide fragment derived from apolipoprotein E (APOE) that competitively binds to low-density lipoprotein receptors modulating associated signaling pathways This compound exhibits both anti-inflammatory and neuroprotective properties with an antagonist activity concentration of 445 nM reported in relevant assays Owing to its specific mechanism of action and demonstrated biological effects COG 133 TFA is widely utilized in research focused on neurodegenerative disorders and inflammation-related pathologies