Learn More
Aviva Systems Biology Corp COG5 Antibody - N-terminal region (ARP50535_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50535_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COG5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human COG5. Alternate names: GOLTC1, GTC90, CDG2IApplicationWB. Species reactivity: Human
Catalog No. 50-014-60370