Learn More
Grainger Dayton Cogged FHP V-Belt, 2L Section, 10 in. Length, 1 ea.
Catalog No. 502770717
Encompass_Preferred
This fractional horsepower (FHP) cogged V-belt is designed for light-duty drive systems, typically operating at 3 HP or less. Constructed with high-performance polyurethane and reinforced with polyester cord, this belt offers excellent durability and resistance to harsh environmental conditions. The cogged design features precision grooves that effectively dissipate heat during operation, while the closed-loop construction ensures consistent performance in HVAC equipment and appliance drive applications.
  • Features a nominal top width of 1/4 in. and a nominal thickness of 5/32 in.
  • Designed for an outside length of 10 in.
  • Material composition provides superior oil and ozone resistance compared to standard neoprene belts.
  • Heat- and stretch-resistant construction supports reliable operation in general purpose applications.
  • Operating temperature range from -22 °F to +194 °F.
Catalog No. 50-277-0717 Supplier Grainger Supplier No. 13V770
This fractional horsepower (FHP) cogged V-belt is designed for light-duty drive systems, typically operating at 3 HP or less. Constructed with high-performance polyurethane and reinforced with polyester cord, this belt offers excellent durability and resistance to harsh environmental conditions. The cogged design features precision grooves that effectively dissipate heat during operation, while the closed-loop construction ensures consistent performance in HVAC equipment and appliance drive applications.
  • Features a nominal top width of 1/4 in. and a nominal thickness of 5/32 in.
  • Designed for an outside length of 10 in.
  • Material composition provides superior oil and ozone resistance compared to standard neoprene belts.
  • Heat- and stretch-resistant construction supports reliable operation in general purpose applications.
  • Operating temperature range from -22 °F to +194 °F.