Learn More
Grainger Dayton FHP V-Belt, Cogged, 12 in Length, 1/4 in Width, 1 ea
Catalog No. 502773005
Encompass_Preferred
This fractional horsepower (FHP) cogged V-belt is designed for use in drive systems typically operating at 3 HP or less, making it suitable for HVAC equipment and appliances. It features a durable polyurethane body reinforced with polyester cords for strength and resistance to heat and stretching.
  • Cogged design with grooves to dissipate heat efficiently
  • Polyurethane construction offers high abrasion resistance and minimal dust production
  • Resistant to oil and ozone exposure
  • Nominal top width of 1/4 in. and thickness of 5/32 in.
  • Operating temperature range of -22°F to 194°F
  • Closed loop, single-sided, raw edge construction
Catalog No. 50-277-3005 Supplier Grainger Supplier No. 13V772
This fractional horsepower (FHP) cogged V-belt is designed for use in drive systems typically operating at 3 HP or less, making it suitable for HVAC equipment and appliances. It features a durable polyurethane body reinforced with polyester cords for strength and resistance to heat and stretching.
  • Cogged design with grooves to dissipate heat efficiently
  • Polyurethane construction offers high abrasion resistance and minimal dust production
  • Resistant to oil and ozone exposure
  • Nominal top width of 1/4 in. and thickness of 5/32 in.
  • Operating temperature range of -22°F to 194°F
  • Closed loop, single-sided, raw edge construction