Learn More
Grainger Dayton Cogged V-Belt, 2L Section, 17 in Length, 1/4 in Top Width, 1 ea
Catalog No. 502770658
Encompass_Preferred
This fractional horsepower (FHP) cogged V-belt is designed for efficient power transmission in machinery and drive systems. The 2L section belt features a closed-loop construction with a polyurethane material composition, offering high abrasion resistance and durability against oil and ozone exposure. Its cogged design includes grooves across the belt width to effectively dissipate heat during operation.
  • Features a 17 in. nominal outside length, 1/4 in. top width, and 5/32 in. thickness
  • Reinforced with polyester cord for heat- and stretch-resistance
  • Designed for temperature ranges from -22°F to 194°F
  • Oil- and heat-resistant construction
  • Static-conductive for safe operation in demanding environments
  • Compatible with single or multiple V-belt drive applications
Catalog No. 50-277-0658 Supplier Grainger Supplier No. 13V777
This fractional horsepower (FHP) cogged V-belt is designed for efficient power transmission in machinery and drive systems. The 2L section belt features a closed-loop construction with a polyurethane material composition, offering high abrasion resistance and durability against oil and ozone exposure. Its cogged design includes grooves across the belt width to effectively dissipate heat during operation.
  • Features a 17 in. nominal outside length, 1/4 in. top width, and 5/32 in. thickness
  • Reinforced with polyester cord for heat- and stretch-resistance
  • Designed for temperature ranges from -22°F to 194°F
  • Oil- and heat-resistant construction
  • Static-conductive for safe operation in demanding environments
  • Compatible with single or multiple V-belt drive applications