Learn More
Grainger Dayton Cogged V-Belt, 3VX Section, 36 in Length, 1 ea.
Catalog No. 502769949
Encompass_Preferred
This 3VX section wedge, cogged V-belt features a raw edge construction for improved power transmission and heat dissipation. The cogged design allows the belt to bend and flex against small-diameter pulleys, making it ideal for drives with twists and turns. It is composed of an abrasion-resistant rubber compound reinforced with high-strength polyester cords.
  • Nominal outside length of 36 in.
  • Top width of 3/8 in. and thickness of 5/16 in.
  • Temperature range of -22°F to 194°F.
  • Resistant to heat, oil, coolants, and ozone exposure.
  • Single-ribbed, wedge profile for enhanced power transfer.
Catalog No. 50-276-9949 Supplier Grainger Supplier No. 13W536
This 3VX section wedge, cogged V-belt features a raw edge construction for improved power transmission and heat dissipation. The cogged design allows the belt to bend and flex against small-diameter pulleys, making it ideal for drives with twists and turns. It is composed of an abrasion-resistant rubber compound reinforced with high-strength polyester cords.
  • Nominal outside length of 36 in.
  • Top width of 3/8 in. and thickness of 5/16 in.
  • Temperature range of -22°F to 194°F.
  • Resistant to heat, oil, coolants, and ozone exposure.
  • Single-ribbed, wedge profile for enhanced power transfer.