Learn More
Grainger Dayton Cogged Classical V-Belt, BX Trade Size, 50 in Length, 1 ea
Catalog No. 502773447
Encompass_Preferred
The cogged, classical V-belt is engineered for performance in single or multiple V-belt drives. Featuring a durable rubber compound body with polyester reinforcing cords and a raw edge construction, this belt offers reliable power transmission and operational stability.
  • Belt trade size BX with an industry number of BX47
  • Outside length of 50 in.
  • Top width of 21/32 in. and thickness of 13/32 in.
  • Operating temperature range from -22°F to 194°F
  • Heat-resistant and oil-resistant properties
  • Cogged design for improved flexibility and heat dissipation
Catalog No. 50-277-3447 Supplier Grainger Supplier No. 13W529
The cogged, classical V-belt is engineered for performance in single or multiple V-belt drives. Featuring a durable rubber compound body with polyester reinforcing cords and a raw edge construction, this belt offers reliable power transmission and operational stability.
  • Belt trade size BX with an industry number of BX47
  • Outside length of 50 in.
  • Top width of 21/32 in. and thickness of 13/32 in.
  • Operating temperature range from -22°F to 194°F
  • Heat-resistant and oil-resistant properties
  • Cogged design for improved flexibility and heat dissipation