Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Cogswell A Elevator - Spade Tip

Catalog No. 21124705 Shop All Surgical Design Products
Change view
Click to view available options
Size:
Adult
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-124-705 Adult
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-705 Supplier Surgical Design™ Supplier No. DELCOGA
Add to Cart
Add to Cart

Cogswell A spade elevator delivers broad, controlled lifting force for elevating teeth or roots during extraction, with durable precision for dental and oral surgery use.

  • Model: Cogswell A Elevator with Spade Tip
  • Broad spade blade ideal for elevating roots and mobilizing teeth
  • Provides excellent leverage for difficult, impacted, or fractured teeth
  • Strong, rigid construction supports heavy-duty use in clinical and forensic settings
  • Ergonomic handle ensures a secure, comfortable grip
  • Optimized blade geometry for improved access in posterior and confined regions
  • Precision-machined from high-grade surgical stainless steel
  • Corrosion- and wear-resistant for repeated sterilization
  • Fully autoclavable and reusable for long-term reliability
  • Applications: General dentistry, oral surgery, forensic dentistry, autopsy and postmortem tooth/mandible access, bone or root elevation

Specifications

Product Type Cogswell A Elevator
Handle Material Stainless Steel
Length (Metric) 15.5 cm
Material Stainless Steel
Size Adult
Autoclavable Autoclavable
Color Silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.