Learn More
Selleck Chemical LLC COH-29 S0283-5MG
SDP
Catalog No. 502173031
Encompass_Preferred
COH29 (RNR Inhibitor COH29) is a potent inhibitor of ribonucleotide reductase (RNR) with anticancer activity COH29 (RNR Inhibitor COH29) inhibits recombinant RNR small subunit (RRM1/RRM2) activity in vitro with IC50 of 16 M
Catalog No. 50-217-3031 Supplier Selleck Chemical LLC Supplier No. S02835MG
COH29 (RNR Inhibitor COH29) is a potent inhibitor of ribonucleotide reductase (RNR) with anticancer activity COH29 (RNR Inhibitor COH29) inhibits recombinant RNR small subunit (RRM1/RRM2) activity in vitro with IC50 of 16 M