Learn More
Medchemexpress LLC Coh-SR4 | 73439-19-7 | 99.5% | 350.03 g/mol | C13H8Cl4N2O | 100MG
SDP
Catalog No. 5000235946
Encompass_Preferred
COH-SR4 is a research-grade small molecule AMPK activator with demonstrated antiproliferative effects in leukemia, melanoma, breast, and lung cancer cell lines. It also inhibits adipocyte differentiation via AMPK activation and is supplied as a solid with documented purity and storage recommendations for laboratory research use only.
  • Activates AMP-activated protein kinase (AMPK) in cellular assays.
  • Demonstrates antiproliferative activity across multiple cancer cell lines.
  • Inhibits adipocyte differentiation, supporting metabolic research.
  • Provided as a solid with documented high purity and storage guidelines.
  • Available in multiple sizes to support varied experimental needs.
  • Intended for laboratory research use only; not for human or veterinary use.
Catalog No. 50-002-35946 Supplier Medchemexpress LLC Supplier No. HY124822100MG
COH-SR4 is a research-grade small molecule AMPK activator with demonstrated antiproliferative effects in leukemia, melanoma, breast, and lung cancer cell lines. It also inhibits adipocyte differentiation via AMPK activation and is supplied as a solid with documented purity and storage recommendations for laboratory research use only.
  • Activates AMP-activated protein kinase (AMPK) in cellular assays.
  • Demonstrates antiproliferative activity across multiple cancer cell lines.
  • Inhibits adipocyte differentiation, supporting metabolic research.
  • Provided as a solid with documented high purity and storage guidelines.
  • Available in multiple sizes to support varied experimental needs.
  • Intended for laboratory research use only; not for human or veterinary use.