Learn More
Medchemexpress LLC COH-SR4 | 73439-19-7 | 99.5% | 350.03 g/mol | C13H8Cl4N2O | 50MG
SDP
Catalog No. 5000237898
Encompass_Preferred
COH-SR4 is a small-molecule AMP-activated protein kinase (AMPK) activator used in preclinical research. It exhibits reported anti-proliferative activity against several cancer cell lines and can inhibit adipocyte differentiation via AMPK activation. Supplied as a high-purity powder for laboratory research and intended for investigational use only.
  • High purity (99.5%) verified by supplier.
  • Known AMPK activation and anti-proliferative activity in vitro.
  • Available in multiple pack sizes to support various study scales.
  • Stable under recommended storage conditions for extended shelf life.
  • Suitable for cancer and metabolic disorder research applications.
Catalog No. 50-002-37898 Supplier Medchemexpress LLC Supplier No. HY12482250MG
COH-SR4 is a small-molecule AMP-activated protein kinase (AMPK) activator used in preclinical research. It exhibits reported anti-proliferative activity against several cancer cell lines and can inhibit adipocyte differentiation via AMPK activation. Supplied as a high-purity powder for laboratory research and intended for investigational use only.
  • High purity (99.5%) verified by supplier.
  • Known AMPK activation and anti-proliferative activity in vitro.
  • Available in multiple pack sizes to support various study scales.
  • Stable under recommended storage conditions for extended shelf life.
  • Suitable for cancer and metabolic disorder research applications.