Learn More
Medchemexpress LLC HY-114304 10mg Medchemexpress, COH000 CAS:1534358-79-6 Purity:>98%
SDP
Catalog No. 501969323
Encompass_Preferred
Medchemexpress, HY-114304 10mg COH000 CAS:1534358-79-6 COH000 is an allosteric, covalent and irreversible inhibitor of ubiquitin-like 1-activating enzyme (SUMO-activating enzyme) (E1), with an IC50 of 0.2 μM for SUMOylation in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9323 Supplier Medchemexpress LLC Supplier No. HY11430410MG
Medchemexpress, HY-114304 10mg COH000 CAS:1534358-79-6 COH000 is an allosteric, covalent and irreversible inhibitor of ubiquitin-like 1-activating enzyme (SUMO-activating enzyme) (E1), with an IC50 of 0.2 μM for SUMOylation in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.