Learn More
Medchemexpress LLC COH29 | 1190932-38-7 | 98.8% | 420.44 | 100 MG
SDP
Catalog No. 5000401648
Encompass_Preferred
COH29 is a potent ribonucleotide reductase (RNR) inhibitor with anticancer activity, effective against various cancer cell lines including ovarian cancer and leukemia. It works by inhibiting the α and β subunits of RNR, and can overcome resistance to other cancer therapies like hydroxyurea and gemcitabine.
  • Potent ribonucleotide reductase (RNR) inhibitor
  • Exhibits anticancer activity
  • Inhibits α and β subunits of RNR
  • Overcomes hydroxyurea and gemcitabine resistance in cancer cells
  • Effectively inhibits proliferation of most cell lines in NCI 60 human cancer panel
  • Little effect on normal fibroblasts or endothelial cells
Catalog No. 50-004-01648 Supplier Medchemexpress LLC Supplier No. HY19931100MG
COH29 is a potent ribonucleotide reductase (RNR) inhibitor with anticancer activity, effective against various cancer cell lines including ovarian cancer and leukemia. It works by inhibiting the α and β subunits of RNR, and can overcome resistance to other cancer therapies like hydroxyurea and gemcitabine.
  • Potent ribonucleotide reductase (RNR) inhibitor
  • Exhibits anticancer activity
  • Inhibits α and β subunits of RNR
  • Overcomes hydroxyurea and gemcitabine resistance in cancer cells
  • Effectively inhibits proliferation of most cell lines in NCI 60 human cancer panel
  • Little effect on normal fibroblasts or endothelial cells