Learn More
Medchemexpress LLC HY-128760 5mg Medchemexpress, COH34 CAS:906439-72-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1287605MG
Encompass_Preferred
Medchemexpress, HY-128760 5mg COH34 CAS:906439-72-3 COH34 is a potent and specific poly(ADP-ribose) glycohydrolase (PARG) inhibitor with an IC50 of 0.37 nM. COH34 binds to the catalytic domain of PARG (Kd=0.547 μM), thereby prolonging PARylation at DNA lesions and trapping DNA repair factors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8928