Learn More
Fine Science Tools Cohan-Vannas Spring Scissors, Stainless Steel, 6 cm
SDP
Catalog No. NC9341133
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 5.7cm/ Serrated: No
Catalog No. NC9341133 Supplier Fine Science Tools Supplier No. 1500002
Add to Cart
Add to Cart
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 5.7cm/ Serrated: No