Learn More
Grainger Danfoss Solenoid Coil, BA024D, 24 V DC, DIN Spade, 45/Pk.
Catalog No. 502763157
Encompass_Preferred
The BA024D solenoid coil is a high-performance electrical component designed for use with solenoid valves, providing reliable continuous-duty operation. This 24 V DC coil features a durable DIN spade connection and is built to withstand varied industrial environments, with an ambient temperature operating range of -40°C to 40°C. It is designed to meet rigorous safety and efficiency standards, including Class H winding insulation for high thermal resistance.
  • Designed for continuous duty operation
  • Includes nut and snap coil fastening
  • Features DIN spade electrical connection (6.3 mm x 0.8 mm)
  • Complies with VDE 0580 coil design standards
  • Class H insulation according to IEC 85
  • IP00 enclosure rating without cable plug
  • Compatible with specified solenoid valve systems
  • Supplied in a multi-pack format
Catalog No. 50-276-3157 Supplier Grainger Supplier No. 161J52
The BA024D solenoid coil is a high-performance electrical component designed for use with solenoid valves, providing reliable continuous-duty operation. This 24 V DC coil features a durable DIN spade connection and is built to withstand varied industrial environments, with an ambient temperature operating range of -40°C to 40°C. It is designed to meet rigorous safety and efficiency standards, including Class H winding insulation for high thermal resistance.
  • Designed for continuous duty operation
  • Includes nut and snap coil fastening
  • Features DIN spade electrical connection (6.3 mm x 0.8 mm)
  • Complies with VDE 0580 coil design standards
  • Class H insulation according to IEC 85
  • IP00 enclosure rating without cable plug
  • Compatible with specified solenoid valve systems
  • Supplied in a multi-pack format