Learn More
Synthware™ Rotary Evaporator Condensers
GREENER_CHOICESDP
Catalog No. 31500820 Shop All Kemtech Products
Change view
Click to view available options
For Use With (Equipment):
Buchi/Diagonal Style Rotary Evaporator
Upright Style Rotary Evaporator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
31-500-820 Buchi/Diagonal Style Rotary Evaporator
31-500-822 Upright Style Rotary Evaporator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-820 Supplier Synthware™ Supplier No. C910200
Add to Cart
Add to Cart
For use with rotary evaporators
C900200:
For vertical style of Heifdorfe.
Accessories:
C900001 Inlet feed tube, 19/38 joint 86.55
RC14 Nylon™ cap 1.15
R201402 Replacement hose connection-straight 1.45
C910200:
Double coil condenser for use with Buchi rotary evaporators. For cooling with tap water in a diagonal position. Has a 19/38 top valve with a 35/20 spherical ball joint.
Accessories:
C900001 Inlet feed tube, 19/38 joint 86.55
RC14 Nylon™ cap 1.15
R201402 Replacement hose connection-straight 1.45
Specifications
Material Borosilicate Glass
Type Condenser
For Use With (Equipment) Buchi/Diagonal Style Rotary Evaporator