Description
- C900200: For vertical style of Heifdorfe.
- Accessories:
- C900001 Inlet feed tube, 19/38 joint 86.55
- RC14 Nylon™ cap 1.15
- R201402 Replacement hose connection-straight 1.45
- C910200: Double coil condenser for use with Buchi rotary evaporators. For cooling with tap water in a diagonal position. Has a 19/38 top valve with a 35/20 spherical ball joint.
- Accessories:
- C900001 Inlet feed tube, 19/38 joint 86.55
- RC14 Nylon™ cap 1.15
- R201402 Replacement hose connection-straight 1.45
Specifications
Specifications
| Material | Borosilicate Glass |
| Type | Condenser |
| For Use With (Equipment) | Upright Style Rotary Evaporator |