Learn More
Chemglass Life Sciences Reactor Capacity: 50L, Flange ID: 300mm, Tubing Diameter: 1/2"
SDP
Catalog No. CG197217
Encompass_Preferred
Reactor Capacity: 50L, Flange ID: 300mm, Tubing Diameter: 1/2". For use with unjacketed cylindrical reaction vessels with a 200mm ID or 300mm ID flange. Coil can be used to cool or heat the contents of a vessel. Fabricated from #304 S.S. tubing with a green PFA coating that is electrically tested to insure complete coverage. This method of manufacture provides for excellent heat transfer and avoids the problems normally associated with heat-shrink PTFE tubing on copper tubing.
Catalog No. CG197217 Supplier Chemglass Life Sciences Supplier No. CG197217
Reactor Capacity: 50L, Flange ID: 300mm, Tubing Diameter: 1/2". For use with unjacketed cylindrical reaction vessels with a 200mm ID or 300mm ID flange. Coil can be used to cool or heat the contents of a vessel. Fabricated from #304 S.S. tubing with a green PFA coating that is electrically tested to insure complete coverage. This method of manufacture provides for excellent heat transfer and avoids the problems normally associated with heat-shrink PTFE tubing on copper tubing.