Learn More
GeminiBio™ GemCell™ Human Serum AB
SDP
Catalog No. 11852135 Shop All Gemini Bio Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
11-852-135 100 mL
1 options
Catalog No. 11-852-135 Supplier GeminiBio™ Supplier No. 100512100
Add to Cart
Add to Cart
GemCell™ Human Serum AB is a cGMP, xeno-free human AB serum from a large pool of healthy U.S. male donors at FDA-licensed facilities. Defibrinated with recombinant human thrombin and non-reactive for viral markers, it supports cell culture, immunology and oncology research.
GemCell™ Human Serum AB is a cGMP, xeno-free human AB serum from a large pool of healthy U.S. male donors at FDA-licensed facilities. Defibrinated with recombinant human thrombin and non-reactive for viral markers, it supports cell culture, immunology and oncology research.
Specifications
Form Serum
Format Bottle
Product Type Human Serum
Quantity 100 mL
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Cell Culture, further Manufacturing, or Research use only. Not for direct Therapeutic use.