Learn More
Kimberly-Clark™ Professional Coil Spring Attachment
Catalog No. 19169103
Click to view available options
KleenGuard™ A10 Bouffant Caps
Catalog No.
19-169-103 KleenGuard™ A10 Bouffant Caps
1 options
This item is not returnable. View return policy
Catalog No. 19-169-103 Supplier Kimberly-Clark™ Professional Supplier No. 14391
This item is not returnable. View return policy
Kimberly-Clark™ Professional Coil Spring Attachment is a locking device to hold a face shield securely in the up or down position.
  • Easily attaches to Jackson Safety Sentry III, Charger and SC-6 Hard Hats and a variety of face shields (all sold separately).
  • Model P adapter is designed to be used with bill-forward facing hats
Specifications
Device Holds 12 in. or 15 1/2 in. visors
Product Type Coil Spring Attachment
For Use With (Equipment) Enables attachment of a variety of Huntsman visors to safety caps