Learn More
MSC Value Collection Coiled Spring Pin, 1/16 in. Diameter, 1 in. Length, 1070/1090 Alloy Steel, 1 ea.
Catalog No. 5000508821
Encompass
This coiled spring pin is designed for versatile fastening applications. Engineered for durability, it provides a secure fit in mechanical assemblies requiring reliable retention.
  • Constructed from 1070/1090 alloy steel
  • Features a 1/16 in. diameter
  • Measures 1 in. in overall length
  • Supplied with a plain finish
  • Designed for inch-based measurement systems
  • Does not meet military specifications
Catalog No. 50-005-08821 Supplier MSC Supplier No. 67501320
This coiled spring pin is designed for versatile fastening applications. Engineered for durability, it provides a secure fit in mechanical assemblies requiring reliable retention.
  • Constructed from 1070/1090 alloy steel
  • Features a 1/16 in. diameter
  • Measures 1 in. in overall length
  • Supplied with a plain finish
  • Designed for inch-based measurement systems
  • Does not meet military specifications