Learn More
Strem, An Ascensus Company CAS 635680-58-9. 250mg. Bis(NN-di-i-propylacetamidinato)cobalt(II) min. 98% (99.99%-Co) PURATREM Co(iPr-MeAMD)2.
SDP
Catalog No. 502110805
Encompass
CAS 635680-58-9. 250mg. Bis(NN-di-i-propylacetamidinato)cobalt(II) min. 98% (99.99%-Co) PURATREM Co(iPr-MeAMD)2. . Molecular Weight 341.40. Molecular Formula C16H34CoN4. Color/form green xtl. Strem 27-0486. http//www.strem.com/catalog/v/27-0486/
Catalog No. 50-211-0805 Supplier Strem, An Ascensus Company Supplier No. 270486250MG
CAS 635680-58-9. 250mg. Bis(NN-di-i-propylacetamidinato)cobalt(II) min. 98% (99.99%-Co) PURATREM Co(iPr-MeAMD)2. . Molecular Weight 341.40. Molecular Formula C16H34CoN4. Color/form green xtl. Strem 27-0486. http//www.strem.com/catalog/v/27-0486/