Learn More
Electron Microscopy Sciences Aurion Col-Aurion Total Protein Stain 50 ML
SDP
Catalog No. 5025029
Encompass_Preferred
Col-Aurion is a colloidal gold particle based total protein stain, developed for the sensitive staining of electrophoretically separated protein bands on nitrocellulose or PVDF blotting membranes. Colloidal gold particles accumulate at the site of the protein bands on the membrane, generating an intense dark red staining pattern. The total protein stain assists in assessing immunoblotting results and to evaluate the effectivity of the blotting procedure.
Storage Col-Aurion has a guaranteed shelf life of 18 months from the date of quality control analysis. Store at 4-8 degree C. Do not freeze.
Catalog No. 50-250-29 Supplier Electron Microscopy Sciences Supplier No. 25536
Col-Aurion is a colloidal gold particle based total protein stain, developed for the sensitive staining of electrophoretically separated protein bands on nitrocellulose or PVDF blotting membranes. Colloidal gold particles accumulate at the site of the protein bands on the membrane, generating an intense dark red staining pattern. The total protein stain assists in assessing immunoblotting results and to evaluate the effectivity of the blotting procedure.
Storage Col-Aurion has a guaranteed shelf life of 18 months from the date of quality control analysis. Store at 4-8 degree C. Do not freeze.