Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 21 mm x 150 mm

Catalog No. NC1661346 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

BioResolve RP mAb Columns feature 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å. These particles are functionalized with a proprietary polyphenyl bonding process for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This product includes Intact mAb Mass Check Standard and mAb Subunit Standard.

  • Designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Operational pH range of 2 to 7.
  • Separation mode: Reversed phase.
  • Particle size: 2.7 μm.
  • Pore size: 450 Å.
  • Dimensions: 21 mm x 150 mm.
  • Recommended flow rate: 0.5 mL/min to 1.0 mL/min.
  • Material: Superficially-porous-silica particles.
  • Applications: Protein analysis, including monoclonal antibodies (mAbs).

Catalog No. NC1661346 Supplier Waters Corp Supplier No. 176004158
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioResolve RP mAb Columns feature 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å. These particles are functionalized with a proprietary polyphenyl bonding process for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This product includes Intact mAb Mass Check Standard and mAb Subunit Standard.

  • Designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Operational pH range of 2 to 7.
  • Separation mode: Reversed phase.
  • Particle size: 2.7 μm.
  • Pore size: 450 Å.
  • Dimensions: 21 mm x 150 mm.
  • Recommended flow rate: 0.5 mL/min to 1.0 mL/min.
  • Material: Superficially-porous-silica particles.
  • Applications: Protein analysis, including monoclonal antibodies (mAbs).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.