Learn More
Perkin Elmer US LLC Elite-1 Capillary Column - 30 m x 0.32 mm I.D. x 1.00 μm
Catalog No. 509046159
Encompass_Preferred
For use at temperatures ranging from -60°C to 320°C/340°C.
Catalog No. 50-904-6159 Supplier Perkin Elmer US LLC Supplier No. N9316024
For use at temperatures ranging from -60°C to 320°C/340°C.