Learn More
Perkin Elmer US LLC Elite-5ms Capillary Column - 30 m x 0.25 mm I.D. x 0.50 µm
Catalog No. 509046393
Encompass_Preferred
For use at temperatures ranging from -60°C to 330°C/350°C.
Catalog No. 50-904-6393 Supplier Perkin Elmer US LLC Supplier No. N9316284
For use at temperatures ranging from -60°C to 330°C/350°C.