Learn More
Perkin Elmer US LLC Elite-5ms Capillary Column - 60 m x 0.25 mm I.D. x 1.00 µm
Catalog No. 509046396
Encompass_Preferred
For use at temperatures ranging from -60°C to 325°C/350°C.
Catalog No. 50-904-6396 Supplier Perkin Elmer US LLC Supplier No. N9316287
For use at temperatures ranging from -60°C to 325°C/350°C.