Learn More
Perkin Elmer US LLC Elite-CLPesticides 2 Capillary Column - 30 m x 0.32 mm I.D. x 0.25 µm
Catalog No. 509046662
Encompass_Preferred
For use at temperatures ranging from -20°C to 240°C/260°C.
Catalog No. 50-904-6662 Supplier Perkin Elmer US LLC Supplier No. N9316670
Add to Cart
Add to Cart
Request a Quote
For use at temperatures ranging from -20°C to 240°C/260°C.