Learn More
Perkin Elmer US LLC Elite-Siltek Deactivated Fused Silica Transfer Line, 5 m x 0.32 mm I.D.
Supplier:  Perkin Elmer US LLC N9316608
Encompass_Preferred
Features and Benefits: Siltek Deactivation process employed for superior inertness, tested to ensure inertness, ideal for the analysis of active samples, maximum Temperature: 380 °C.
Catalog No. 50-904-6600