Learn More
Perkin Elmer US LLC Elite-WAX Capillary Column - 30 m x 0.53 mm I.D. x 1.00 µm
Catalog No. 509046470
Encompass_Preferred
For use at temperatures ranging from 20°C to 240°C/250°C.
Catalog No. 50-904-6470 Supplier Perkin Elmer US LLC Supplier No. N9316427
For use at temperatures ranging from 20°C to 240°C/250°C.